Aluminium hydroxide ≥99.9% (metals basis)

Supplier: Aladdin Scientific

A137846-250G
ALA137846-250GEA 30.31 USD
ALA137846-250G
Aluminium hydroxide ≥99.9% (metals basis)
Aluminium hydroxide
Formula: Al(OH)₃
Melting Pt: 300°C
Density: 2.4 g/cm³
MDL Number: MFCD00003420
CAS Number: 21645-51-2
EINECS: 244-492-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR