Aniracetam ≥98% (by GC), Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(4-Methoxybenzoyl)pyrrolidin-2-one, 1-(4-Methoxybenzoyl)-2-pyrrolidone

A137017-1G
ALA137017-1GEA 80.66 USD
ALA137017-1G
Aniracetam ≥98% (by GC), Moligand™
Aniracetam

Aniracetam has modulatory actions through allosteric potentiation of reduction of glutamate receptor desensitization, AMPA specific receptors, and potentiation of metabotropic glutamate receptor activity. AMPA is a transmembrane glutamate receptor composed of the subunits GluR-1,2,3,4.A nootropic agent with modulatory actions at Glutamate and AMPA receptors demonstrating anxiolytic activity.

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00153767
CAS Number: 72432-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59725144b50b289f1c2606cc337b0d65ffd1561b&vid=59725144b50b289f1c2606cc337b0d65ffd1561b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR