5-Aminoisoquinoline dihydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A136406-250MG
ALA136406250MGEA 379.91 USD
ALA136406250MG
5-Aminoisoquinoline dihydrochloride ≥97%
5-Aminoisoquinoline dihydrochloride
Formula: C₉H₉ClN₂
MDL Number: MFCD00035269
CAS Number: 58143-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d8613bb93ad28273d94911482fa485c7ee509d&vid=f2d8613bb93ad28273d94911482fa485c7ee509d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR