4'-Acetoxyacetanilide, pharma grade

Supplier: ALADDIN SCIENTIFIC
Warning

A135410-100MG
ALA135410100MGEA 131.96 USD
ALA135410100MG
4'-Acetoxyacetanilide, pharma grade
4'-Acetoxyacetanilide
Formula: C₁₀H₁₁NO₃
Melting Pt: 155 °C
MDL Number: MFCD00059205
CAS Number: 2623-33-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a50a9df2fdf6bcad31127b823c697deec72aea1&vid=3a50a9df2fdf6bcad31127b823c697deec72aea1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR