9,10-Anthracenediyl-bis(methylene)dimalonic acid ≥90%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2,2'-(Anthracene-9,10-diylbis(methylene))dimalonic acid

A131481-100MG A131481-10MG A131481-1G A131481-25MG A131481-500MG A131481-50MG
ALA131481100MGEA 187.06 USD
ALA131481100MG ALA131481-10MG ALA131481-1G ALA131481-25MG ALA131481500MG ALA131481-50MG
9,10-Anthracenediyl-bis(methylene)dimalonic acid ≥90%
9,10-Anthracenediyl-bis(methylene)dimalonic acid

Product Describtion:Reagent for the assay of singlett oxygen; it has better characteristics than 9,10-anthracenediyl-bis-dipropionic acid; In addition to the emission maximum at 407 nm, there are lower maxima at 431, 457 and 485 nmA useful O2 determining agent.Product Application:9,10-Anthracenediyl-bis(methylene)dimalonic acid is widely used as a singlet oxygen detector probe. 9,10-Anthracenediyl-bis(methylene)dimalonic acid is suitable for assessing the photodynamic effect of curcumin Vibrio parahaemolyticus, which is a significant cause of bacterial diarrhea

Formula: C₂₂H₁₈O₈
Storage Temperature: Ambient
MDL Number: MFCD03095464
CAS Number: 307554-62-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f99c7072f32f388928fee84eb5185750751153e0&vid=f99c7072f32f388928fee84eb5185750751153e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR