AMG-458 ≥99%
Supplier: ALADDIN SCIENTIFIC
A129793-25MG
A129793-50MG
A129793-5MG
A129793-10MG
ALA129793-25MGEA
554.71
USD
ALA129793-25MG
ALA129793-50MG
ALA129793-5MG
ALA129793-10MG
AMG-458 ≥99%
AMG-458
Product Describtion:AMG-458 is a potent, selective and orally bioavailable c-Met inhibitor, with Ki values of 1.2 nM and 2.0 nM for human and mouse c-Met, respectively.
Formula:
C₃₀H₂₉N₅O₅ Storage Temperature: Freezer |
CAS Number:
913376-83-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...