AMG-458 ≥99%

Supplier: ALADDIN SCIENTIFIC

A129793-25MG A129793-50MG A129793-5MG A129793-10MG
ALA129793-25MGEA 554.71 USD
ALA129793-25MG ALA129793-50MG ALA129793-5MG ALA129793-10MG
AMG-458 ≥99%
AMG-458

Product Describtion:AMG-458 is a potent, selective and orally bioavailable c-Met inhibitor, with Ki values of 1.2 nM and 2.0 nM for human and mouse c-Met, respectively.

Formula: C₃₀H₂₉N₅O₅
Storage Temperature: Freezer
CAS Number: 913376-83-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46baf5364ce691c4af45c6406b33859a8d6f84d5&vid=46baf5364ce691c4af45c6406b33859a8d6f84d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR