AS-703026 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC

Synonyms: (S)-N-(2,3-Dihydroxypropyl)-3-((2-fluoro-4-iodophenyl)amino)isonicotinamide, MSC1936369B, Pimasertib

A125227-50MG A125227-5MG
ALA125227-50MGEA 186.11 USD
ALA125227-50MG ALA125227-5MG
AS-703026 ≥98%, Moligand™
AS-703026

Pimasertib (AS-703026) is a highly selective, potent, ATP non-competitive allosteric inhibitor of MEK1/2 with IC50 of 5 nM-2 μM in MM cell lines.A potent inhibitor of MEK1 and MEK2.

Formula: C₁₅H₁₅FIN₃O₃
Storage Temperature: Freezer
MDL Number: MFCD16660676
CAS Number: 1236699-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d59b08c1f3354564891dc85ff478a427289dfef9&vid=d59b08c1f3354564891dc85ff478a427289dfef9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR