2-Aminopyridin-4-ol ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Amino-4-hydroxypyridine

A124105-1G A124105-25G A124105-5G
ALA124105-1GEA 29.36 USD
ALA124105-1G ALA124105-25G ALA124105-5G
2-Aminopyridin-4-ol ≥97% (by HPLC)
2-Aminopyridin-4-ol
Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD04114161
CAS Number: 33631-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=803ef7ad0b490528e9b9a07cad529aad44e949c8&vid=803ef7ad0b490528e9b9a07cad529aad44e949c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR