5-Amino-1H-imidazole-4-carboxamide hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A124057-5G
ALA124057-5GEA 62.61 USD
ALA124057-5G
5-Amino-1H-imidazole-4-carboxamide hydrochloride ≥98%
5-Amino-1H-imidazole-4-carboxamide hydrochloride

Starting material for synthesis of the antitumour agent temozolomide合成杂环化合物(例如嘌呤和嘧啶)的反应物。

Formula: C₄H₇ClN₄O
Melting Pt: 270°C
Storage Temperature: Ambient
MDL Number: MFCD00012704
CAS Number: 72-40-2
EINECS: 200-778-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ac3cfb7b6838db4c76f3b237af5e74d053ff177&vid=5ac3cfb7b6838db4c76f3b237af5e74d053ff177&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR