4-Amino-3-iodobenzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A123913-1G
ALA123913-1GEA 121.51 USD
ALA123913-1G
4-Amino-3-iodobenzoic acid ≥98%
4-Amino-3-iodobenzoic acid
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Melting Pt: 208 °C
Storage Temperature: Ambient
MDL Number: MFCD01660550
CAS Number: 2122-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c63b2a75b5c464b71d24b2b11bfe3dde0843c6ee&vid=c63b2a75b5c464b71d24b2b11bfe3dde0843c6ee&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR