4-Amino-α,α,α-trifluoro-o-toluic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A123911-100G A123911-1G A123911-25G A123911-5G
ALA123911-100GEA 2017.71 USD
ALA123911-100G ALA123911-1G ALA123911-25G ALA123911-5G
4-Amino-α,α,α-trifluoro-o-toluic acid ≥97%
4-Amino-α,α,α-trifluoro-o-toluic acid
Formula: C₈H₆F₃NO₂
Melting Pt: 183-186°C
MDL Number: MFCD03407959
CAS Number: 393-06-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1125e7169adc85e59f2ff12a433410ab9530adc1&vid=1125e7169adc85e59f2ff12a433410ab9530adc1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR