Isoluminol (4-aminophthalhydrazide) ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A120790-1G
ALA120790-1GEA 79.71 USD
ALA120790-1G
Isoluminol (4-aminophthalhydrazide) ≥98%
Isoluminol (4-Aminophthalhydrazide)

Produces chemiluminescence

Formula: C₈H₇N₃O₂
Melting Pt: 300°C
MDL Number: MFCD00010560
CAS Number: 3682-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d10d4b8cb84b05be8194a0d9e48da64c777a14f&vid=1d10d4b8cb84b05be8194a0d9e48da64c777a14f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR