L(+)-Asparagine monohydrate ≥99% for cell culture animal-free

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O

A119452-100G
ALA119452-100GEA 86.36 USD
ALA119452-100G
L(+)-Asparagine monohydrate ≥99% for cell culture animal-free
L(+)-Asparagine monohydrate
Formula: C₄H₈N₂O₃·1H₂O
MW: 150.13 g/mol
Melting Pt: 233-235°C
Density: 1.541 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00151038
CAS Number: 5794-13-8
EINECS: 200-735-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc1ccb6b1951c3d14646acb7f6ef7a4994db4215&vid=bc1ccb6b1951c3d14646acb7f6ef7a4994db4215&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR