Acetonitrile ≥99.5% ACS

Supplier: ALADDIN SCIENTIFIC
Danger

A119416-500ML
ALA119416500MLEA 37.91 USD
ALA119416500ML
Acetonitrile ≥99.5% ACS
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81-82°C
Melting Pt: -46°C
Density: 0.781 g/cm³
Flash Pt: 2 °C
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR