Aluminium oxide, α-phase nanoparticles 20% (w/w) in water 30 nm

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: ɑ-Tonerde, α-Alumina

A119405-100ML
ALA119405100MLEA 52.16 USD
ALA119405100ML
Aluminium oxide, α-phase nanoparticles 20% (w/w) in water 30 nm
Aluminium oxide, α-phase
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2977°C
Melting Pt: 2000°C
Density: 1.178 g/cm³
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=309bc68ced5c1603331c75bfb8899e39246fc18c&vid=309bc68ced5c1603331c75bfb8899e39246fc18c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR