Ammonium heptamolybdate tetrahydrate ≥99.9% (metals basis)

Supplier: Aladdin Scientific
Warning

A116378-100G
ALA116378-100GEA 151.9 USD
ALA116378-100G
Ammonium heptamolybdate tetrahydrate ≥99.9% (metals basis)
Ammonium heptamolybdate tetrahydrate

Ammonium molybdate tetrahydrate is used in cryo-negative staining and helps visualize particles while preserving the sample. Cryo-negative staining with Ammonium molybdate has been used to visualize Escherichia coli chaperone GroEL, 20S proteasome from moss, tobacco mosaic virus, bacateriophage T2, tomato bushy stunt virus (TBSV), bovine liver catalase crystals, turnip yellow mosaic virus and keyhole limpet hemocyanin (KLH) TYPES 1 and 2. Ammonium molybdate tetrahydrate has been used to catalytically desulfurize thiophene and its derivatives.

Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Melting Pt: 190°C
Density: 2.498 g/cm³
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-320-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR