Glycine ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

A110749-500G
ALA110749-500GEA 25.9 USD
ALA110749-500G
Glycine ≥99%, Moligand™
Glycine

Glycine is widely used as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers (Tris-Glycine) where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. Glycine (100 mM, pH 2.0-2.5) is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin′s transfer buffer for Western blots

Formula: NH₂CH₂COOH
Melting Pt: 240 °C
Density: 1.03 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4673b851136f8e62756909336b4e4c17284f242e&vid=4673b851136f8e62756909336b4e4c17284f242e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR