4-Aminobutyric acid ≥99%, Moligand™

Supplier: Aladdin Scientific
Warning

A104200-25G
ALA104200-25GEA 22.9 USD
ALA104200-25G
4-Aminobutyric acid ≥99%, Moligand™
4-Aminobutyric acid
Formula: C₄H₉NO₂
Melting Pt: 195°C
Density: 1.11 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00008226
CAS Number: 56-12-2
EINECS: 200-258-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR