Ammonium iodide ≥99% AR

Supplier: ALADDIN SCIENTIFIC

A103671-500G
ALA103671-500GEA 140.9 USD
ALA103671-500G
Ammonium iodide ≥99% AR
Ammonium iodide
Formula: NH₄I
Density: 2.514 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR