L(+)-Ascorbic acid ≥99.99% (metals basis), PrimorTrace™

Supplier: ALADDIN SCIENTIFIC

A103534-100G
ALA103534-100GEA 39.9 USD
ALA103534-100G
L(+)-Ascorbic acid ≥99.99% (metals basis), PrimorTrace™
L(+)-Ascorbic acid

A calcium channel inhibitor.

Formula: C₆H₈O₆
Melting Pt: 190-192°C
Density: 1.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
EINECS: 200-066-2, 200-066-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bce0e40507fdcf7f8c383ef0d10aaf7cb95cd38&vid=8bce0e40507fdcf7f8c383ef0d10aaf7cb95cd38&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR