(3-Aminopropyl)trimethoxysilane ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-(Trimethoxysilyl)propylamine, 3-(Trimethoxysilyl)propan-1-amine, Trimethoxy(3-aminopropyl)silane, 3-Aminopropyltrimethoxysilane

A100943-500G
ALA100943-500GEA 116.9 USD
ALA100943-500G
(3-Aminopropyl)trimethoxysilane ≥97%
(3-Aminopropyl)trimethoxysilane

(3-Aminopropyl) trimethoxysilane (APTMS) is an aminosilane which is used in silanization processes as a silane coupling agent. It functionalizes substrates with alkoxysilane molecules. (3-Aminopropyl)trimethoxysilane(APTMS) is an amino-silane that is mainly used as a silane coupling agent for the surface modification of a variety of nanomaterials. APTMS based films form a thermally stable layer on different substratesIt can be multilayered on SiOx substrates by a layer by layer self assembly,and also used as a silane coupling agent on silver nanoparticles.

Formula: H₂NCH₂CH₂CH₂Si(OCH₃)₃
MW: 179.29 g/mol
Boiling Pt: 194
Flash Pt: 86°C
Storage Temperature: Ambient
MDL Number: MFCD00008206
CAS Number: 13822-56-5
EINECS: 237-511-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a46f884e19d69bfa8b8db7857cf4fec1d25be7ec&vid=a46f884e19d69bfa8b8db7857cf4fec1d25be7ec&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR