Acetylthiocholine iodide ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A100869-25G
ALA100869-25GEA 263.9 USD
ALA100869-25G
Acetylthiocholine iodide ≥98%
Acetylthiocholine iodide

Acetylthiocholine iodide is an AChE (acetylcholinesterase) substrate and AChR (nicotinic acetylcholine receptor)agonist.A nAChR agonist and substrate of acetylcholinesterase

Formula: H₃CC(S)O(CH₂)₂N(CH₃)₃I
Melting Pt: 205-210°C
Storage Temperature: Refrigerator
MDL Number: MFCD00011819
CAS Number: 1866-15-5
EINECS: 217-474-1
UN: 2811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=033a006f7627f0cc74af946815a23691eded1ed2&vid=033a006f7627f0cc74af946815a23691eded1ed2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR