AMP (2-Amino-2-methylpropanol) ≥90%

Supplier: ALADDIN SCIENTIFIC
Danger

A100592-100ML
ALA100592-100MLEA 19.9 USD
ALA100592-100ML
AMP (2-Amino-2-methylpropanol) ≥90%
AMP (2-Amino-2-methylpropanol)

Used in the preparation of buffer solutions, suitable for the determination of alkaline phosphatase.2-Amino-2-methyl-1-propanol can be used to study biological buffers. 2-Amino-2-methyl-1-propanol has been used in a study to report on an ATR-FTIR spectroscopic investigation of the CO(2) absorption characteristics of a series of heterocyclic diamines. 2-Amino-2-methyl-1-propanol has also been used in a study to develop a mild and convenient method for preparing 4,4-dimethyloxazoline (DMOX) derivatives of fatty acids for GC-MS analysis.

Formula: (CH₃)₂C(NH₂)CH₂OH
Boiling Pt: 165℃
Melting Pt: 24-28°C
Density: 0.934 g/cm³
Flash Pt: 67°C
MDL Number: MFCD00008051
CAS Number: 124-68-5
EINECS: 204-709-8
UN: 1993
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b7508ad906e0f7c86b18e23e5ad4859a17a9e3d&vid=6b7508ad906e0f7c86b18e23e5ad4859a17a9e3d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR