Ammonium formate ≥99.99% (metals basis), PrimorTrace™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Formic acid ammonium salt

A100184-500G
ALA100184-500GEA 200.9 USD
ALA100184-500G
Ammonium formate ≥99.99% (metals basis), PrimorTrace™
Ammonium formate

Ammoniumformate is a derivative of formic acid widely used as a hydrogen transfer reagentand formylating agent. It is also used in the preparation and storage of formicacid.Application:Ammonium formate can be used as: Hydrogen source in reduction of aromatic nitro compounds to corresponding amino derivatives in presence of Cu nanoparticles as catalyst. Formylating agent for synthesis of N-formyl amino acid esters. An OH radical scavenger in the preparation of nanocrystalline uranium and/or thorium oxides.

Formula: HCO₂NH₄
MW: 63.06 g/mol
Melting Pt: 116°C
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f621766450d8bc681cc4bcf82b4956359b418d5b&vid=f621766450d8bc681cc4bcf82b4956359b418d5b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR