CrossLab Gas Tight Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
SG003987
AGSG003987EA 290 USD
AGSG003987
CrossLab Gas Tight Syringes, Agilent Technologies
Syringes Syringes for Chromatography
Gas tight syringes for dispensing gases and liquids.
  • Precisely made for optimal and repeatable performance through accurate volume delivery
These gas tight syringes are constructed of borosilicate glass and PTFE plunger tips. An interference fit between the plunger tip and the barrel enhances the gas sealing properties of the syringe.