Torch Supplies for MP-AES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
G8003-68003
AGG8003-68003EA 153 USD
AGG8003-68003
Torch Supplies for MP-AES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Torch clamps, torch cleaning stand, and inert PTFE ball joint adapter for the MP-AES.
  • Torch cleaning stand for the MP-AES torch: Provides a stable platform for suspending the outer tube, intermediate tube, and injector in the acid cleaning solution
  • Encloses the container for the acid cleaning solution, reducing the chance of accidental spillage
  • Prevents damage to the fragile end of the quartz outer tube of the torch from contact with the base of the container used for the acid cleaning solution
  • Reduces premature degradation of the elastomer materials on the torch body as the seal between the torch body and the torch plate limits exposure to acid fumes from the cleaning solution
Agilent MP-AES torch supplies include torch clamps, torch cleaning stand, and PTFE ball joint adaptor for inert MP-AES torch used with the MP-AES instruments.