Thermal Conductivity Detector (TCD), Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
G3432-60220
AGG3432-60220EA 3805 USD
AGG3432-60220
Thermal Conductivity Detector (TCD), Agilent Technologies
Chromatography Detectors
Detector responds to all compounds and compares the thermal conductivities of two gas flows using a single-filament, single-column design.
  • Designed, tested and manufactured by Agilent engineers to work perfectly with our instruments Superior performance
The thermal conductivity detector responds to all compounds, excluding carrier gas. The TCD compares the thermal conductivities of two gas flows – pure carrier gas (also called the reference gas) and carrier gas plus sample components (also called column effluent), using a single-filament, single-column design.