CrossLab ICP-MS Calibration Standards, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
G3280-60590
AGG3280-60590KT 1087 USD
AGG3280-60590
CrossLab ICP-MS Calibration Standards, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Single-element and multi-element reference materials for ICP-MS.
  • Highest quality and designed, developed, and manufactured to guarantee at least one year of shelf life
To ensure the stability and performance of ICP-MS analyses, Agilent offers a comprehensive portfolio of single-element and multi-element reference materials that test the efficacy of the instrument.
With the assurance of NIST traceability, Agilent's selection of standards offers accurate, reliable calibrations at great value. All starting materials have a purity of 99.999% whenever possible.