Hydride Generation Supplies for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
G3138-65128
AGG3138-65128PK 249.3 USD
AGG3138-65128
Hydride Generation Supplies for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
ISIS Hydride Generation Accessories for analysis of As, Sb, Se, Sn and Hg with ICP-MS systems.
  • ISIS hydride generation accessories are specially designed for Agilent instruments to ensure high quality results
  • Flexible selection of accessories for every instrument configuration
  • Convenient kits and accessories are simple to order and make user maintenance easy to schedule and execute
  • Hydride generation offers the best sensitivity for As, Sb, Se, Sn and Hg with ICP-MS, which are all hydride forming elements
Agilent manufactures a range of high quality ICP-MS replacements and consumables including ISIS hydride generation accessories. The range of accessories include PFA, PTFE and pump tubes of various lengths, union and cross joints, a connector assembly with a range of connector options and several additional fixtures and fittings. The ISIS hydride generation accessory is designed to demanding specifications and delivers the market-leading ICP-MS performance that you have come to expect. Maintain system performance and ensure that consumables are periodically replaced in accordance with recommended maintenance schedules.