Bond Elut PBA Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
A496121C
AGA496121CEA 735 USD
AGA496121C
Bond Elut PBA Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut PBA uses phenylboronic acid functionality to retain analytes.
  • Normal phase or anion-exchange sorbent
  • Weaker anion exchange than SAX
  • Amenable to separating structural isomers
Bond Elut PBA is a unique silica SPE sorbent containing a phenylboronic acid functionality that can retain analytes through a reversible covalent bond. This strong covalent retention mechanism enables high specificity and cleanliness. The boronate group has a strong affinity for cis-diol-containing compounds, such as catechols and nucleic acids.