iTLC-SA, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
A120B12
103579-620PK 607.81 USD
103579-620 AGA120B12
iTLC-SA, Agilent Technologies
Filters Filter Paper Chromatography Paper
This SA (contains sodium salt) chromatography paper is used in thin layer chromatography applications such as evaluating radioisotope purity.
  • More convenient with faster developing times than traditional thin layer chromatography plates
  • Open porous matrix provides fast developing time (often less than 20 min)
  • Can also be used to separate lipids and other non-polar compounds
  • Sample elution is easy and without interference from organic binders
  • Can be cut with scissors or a cutter
Instant Thin Layer Chromatography (iTLC) Medium is a binderless, glass microfiber chromatography paper or sheet impregnated with a silicic acid (SA). It is a porous, slightly acidic sheet that provides excellent resolution.