CrossLab Tubing for Cary UV Accessories, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
9910052900
AG9910052900KT 236.2 USD
AG9910052900
CrossLab Tubing for Cary UV Accessories, Agilent Technologies
Tubing Chromatography Tubing
Tubing spares and kits for Cary UV-Vis spectrophotometer instruments and accessories.
  • Designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
These allow you to replace peristaltic pump tubing and inlet/outlet connections for flow cells to help you keep your system operating at peak performance.