Spray Chambers for PerkinElmer
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
8003-0523
102971-914EA 730.28 USD
102971-914 102971-916 102972-296 102972-298 102972-300 102972-460 102972-464 102972-470 102972-472 102972-474 102972-476 102972-478 102972-480 102972-482 102972-484 102972-486 102972-556 102972-570 102972-572 AG8003-0523
Spray Chambers for PerkinElmer
Chromatography Supplies Chromatography Spare Parts
These spray chambers and supplies are fully compatible with your PerkinElmer ICP-MS.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout. Alternatively, the double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.