Spray Chambers and Supplies for PerkinElmer
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
8003-0463
102971-872EA 2248.12 USD
102971-872 102971-874 102971-876 102971-994 102972-310 102972-410 102972-416 102972-418 102977-372 102977-374 102995-188 102995-202 102995-224 AG8003-0463
Spray Chambers and Supplies for PerkinElmer
Chromatography Supplies Chromatography Spare Parts
These spray chambers and supplies are fully compatible with your PerkinElmer ICP-OES.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout. Alternatively, the double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.