In-Line Filters for Thermo/Dionex LC Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
8002-0806
AG8002-0806EA 116.4 USD
AG8002-0806
In-Line Filters for Thermo/Dionex LC Systems, Agilent Technologies
Filters In-Line Filters
High-quality in-line filters for selected Thermo Fisher Scientific HPLC systems.
  • In-line solvent filters, installed between the LC pump and injector, prevent blockages by removing particles from solvents before they reach the injector
  • Column inlet filters, positioned immediately before an LC column, remove particles from both the injection system and sample
Agilent offers in-line filters for Thermo Fisher Scientific HPLC systems, made from clean, high-quality stainless steel parts. These filters are designed to reduce column inlet frit contamination and blockage, which can increase backpressure and reduce efficiency, especially in microbore columns.