Bond Elut PSA Plates
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7542101C
101473-750EA 406.71 USD
101473-750 AG7542101C
Bond Elut PSA Plates
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut PSA is an alkylated amine with two different amino functionalities and slightly higher pKa and ionic capacity than Bond Elut NH₂ for polar compounds.
  • Alternative choice to Bond Elut NH₂ for polar compounds
  • Higher ionic capacity than NH₂
It is an alkylated amine sorbent that contains two different amino functionalities, one secondary and one primary. This gives a slightly higher pKa and ionic capacity compared to Bond Elut NH₂. PSA has a significantly higher carbon load than most amino functional sorbents and thus is a better choice for polar compounds, which retain too strongly on Bond Elut NH₂.