Bond Elut Certify II Plate
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7541801C
101473-746EA 384.24 USD
101473-746 AG7541801C
Bond Elut Certify II Plate
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut Certify II is an anion exchange/mixed-mode SPE product suitable for extraction of acidic compounds from samples.
  • Ideal for nonpolar and anionic compounds
  • Optimized for acidic drug analysis
  • Bimodal, nonpolar and strong anion exchange
Bond Elut Certify II was developed specifically for the rapid and effective extraction of acidic drugs and metabolites from urine and other biological matrices. Certify II is a mixed-mode cartridge packed with nonpolar C8 and strong anion-exchange (SAX) sorbent.
It has been optimized for acidic drugs, such as 11-nor-delta-9-tetrahydrocannibinol-carboxylic acid, salicylic acid, ibuprofen, acetaminophen, and other compounds that possess both nonpolar and anionic characteristics. For forensic use.