Bond Elut DEA Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7541701C
AG7541701CEA 513 USD
AG7541701C
Bond Elut DEA Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut DEA is ideal for aqueous anion-exchange or as a polar sorbent with nonpolar matrices.
  • Weak anion exchanger
  • More polar than C8 but less polar than C2 or CN-E
  • Alkyl side chains exhibit moderately nonpolar characteristics
Bond Elut DEA (diethylaminopropyl) is similar to Bond Elut NH2, but with a slightly lower capacity as an anion-exchange sorbent. DEA has a moderately nonpolar character due to the alkyl side chains on the amino functionality. These groups still afford a medium level of polarity, higher than C8, but less polar than C2 or CN-E.