Bond Elut SI Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7541301C
101473-732EA 359.52 USD
101473-732 101473-756 AG7541301C
Bond Elut SI Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut SI is particularly effective at separating compounds with a very similar structure.
  • Highly polar phase retains polar molecules from nonpolar matrices
  • High purity silica
  • Separate compounds with very similar structures
Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut SI is particularly effective at separating compounds with a very similar structure. Applying the analytes in a non-polar solvent, then increasing the solvent polarityMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable