Bond Elut Certify Plates
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7540901C
101402-232EA 562.87 USD
101402-232 101402-234 101402-236 101473-724 101473-832 101473-834 103579-950 AG7540901C AG75409050
Bond Elut Certify Plates
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut certify delivers a cation exchange/mixed-mode SPE approach that is ideal for extraction of drugs from urine and blood, along with many other applications.
  • Special mixed-mode sorbent bed
  • Broad application range for aqueous extraction
  • Bimodal, nonpolar, and strong cation exchange
Bond Elut certify extraction cartridge utilizes a packed bed consisting of a special, nonpolar C8 sorbent and a strong cation exchanger (SCX). Certify is most commonly used to extract basic (cationic) drugs from urine and blood, but it is also very effective for extracting a wide range of compounds from a diverse number of aqueous matrices.