Well Plate Tubes for Bond Elut PPL Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7531901C
AG7531901CPK 105 USD
AG7531901C
Well Plate Tubes for Bond Elut PPL Plates, Agilent Technologies
Tubes Test Tubes
Bond Elut PPL is a modified styrene-divinylbenzene polymer that retains polar analytes.
  • Modified styrene-divinylbenzene polymer
  • Large particle size enables fast extraction speeds
  • High surface area and capacity for polar analytes
Bond Elut PPL (Priority PolLutant) is a styrene-divinylbenzene (SDVB) polymer modified with a proprietary nonpolar surface. PPL will retain even the most polar classes of analytes. The large particle size eases flow for viscous or particulate-rich water samples, while the large surface area and strong hydrophobicity ensure reproducible extractions with high recoveries.