QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
QuEChERS dispersive solid phase extraction (SPE) kits supplied in 15 ml centrifuge tubes accommodate the aliquot volumes specified by current AOAC and EN methodologies.
- Non hazardous
The sorbent extracts the sample matrix, while MgSO₄ helps remove excess water and improve analyte partitioning.