CrossLab Solvent Filtration and Filter Membranes for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5191-4337 5191-4338 5191-4341 5191-6776 5191-4336 5191-4339 5191-4340 5191-6778
AG5191-4337PK 272.4 USD
AG5191-4337 AG5191-4338 AG5191-4341 AG5191-6776 AG5191-4336 AG5191-4339 AG5191-4340 AG5191-6778
CrossLab Solvent Filtration and Filter Membranes for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Improve LC system performance by removing contaminants and degassing the mobile phase.
  • Degas eluents as particulates are removed
  • Prevent the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph
  • Eliminate pump downtime caused by air locks and particulates in check valves
  • Decrease piston wear
  • Increase column life
  • Increase solvent inlet lifetime
Mobile phase filters can improve the performance of your LC system by removing contaminants from solvents and degassing solvents in systems lacking on-line degassers.
Choose from a variety of different membrane filters compatible with normal- and reversed-phase mobile phases.