CrossLab CE and CE/MS Tools, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5183-4669
AG5183-4669EA 646.9 USD
AG5183-4669
CrossLab CE and CE/MS Tools, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
For fast, convenient, and reproducible preparation of detection windows on fused silica capillaries.
  • Reproducible preparation of detection windows on fused-silica capillaries
  • Polyimide coating is removed without compromising the inner polymeric coating
  • Tool features three glass blocks with fine grooves to control the size of windows
Agilent supplies CE tools such as the window etching tool, designed for fast, convenient, and reproducible preparation of detection windows on fused-silica capillaries. The polyimide coating is removed without destroying the inner polymeric coating. The tool contains three glass blocks with fine grooves, precisely controlling the size of the windows.