GC Autosampler Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5181-1267 5181-3354 5181-3360 5181-8810 G4513-80206 G4513-80209 5181-1273 5181-3361 5182-0836 5182-0875 5183-4730 8010-0163 8010-0265 8010-0357 9301-0725 G4513-80203 G4513-80204 G4513-80208 G4513-80213 G4513-80220 G4513-80226 5181-3321 5181-8809 5182-0835 5183-0316 5183-2042 5183-4728 5183-4729 5188-5246 5188-5247 8004-0001 8010-0164 8010-0351 8010-0364 G4513-80200 G4513-80201 G4513-80202 G4513-80205 G4513-80215 G4513-80216 G4513-80229 G4513-80234 G4513-80235 G4513-80243 G4513-80244 G6500-80109 G6500-80116 5181-3356 5181-8806 5181-8813 5182-0834 5182-9799 5183-0314 8010-0118 8010-0129 8010-0130 8010-0168 8010-0174 8010-0270 8010-0352 8010-0355 8010-0356 8010-0358 8010-0363 8010-0371 9301-0714 G4200-80119 G4513-60560 G4513-60561 G4513-80210 G4513-80211 G4513-80212 G4513-80218 G4513-80221 G4513-80222 G4513-80241 G4513-80242 G6500-80107 G6500-80114 G6500-80115 5182-9622 5182-9626 5182-9734 5188-6485 5190-3984 8010-0090 8010-0138 8010-0169 8010-0173 8010-0184 8010-0197 8010-0266 8010-0271 8010-0353 8010-0360 8010-0361 8010-0362 G4200-80118 G4513-80207 G4513-80219 G4513-80223 G4513-80224 G4513-80228 G4513-80230 G4513-80233 G6500-80111 G6500-80113
AG5181-1267EA 72 USD
AG5181-1267 AG5181-3354 AG5181-3360 AG5181-8810 AGG4513-80206 AGG4513-80209 AG5181-1273 AG5181-3361 AG5182-0836 AG5182-0875 AG5183-4730 AG8010-0163 AG8010-0265 AG8010-0357 AG9301-0725 AGG4513-80203 AGG4513-80204 AGG4513-80208 AGG4513-80213 AGG4513-80220 AGG4513-80226 AG5181-3321 AG5181-8809 AG5182-0835 AG5183-0316 AG5183-2042 AG5183-4728 AG5183-4729 AG5188-5246 AG5188-5247 AG8004-0001 AG8010-0164 AG8010-0351 AG8010-0364 AGG4513-80200 AGG4513-80201 AGG4513-80202 AGG4513-80205 AGG4513-80215 AGG4513-80216 AGG4513-80229 AGG4513-80234 AGG4513-80235 AGG4513-80243 AGG4513-80244 AGG6500-80109 AGG6500-80116 AG5181-3356 AG5181-8806 AG5181-8813 AG5182-0834 AG5182-9799 AG5183-0314 AG8010-0118 AG8010-0129 AG8010-0130 AG8010-0168 AG8010-0174 AG8010-0270 AG8010-0352 AG8010-0355 AG8010-0356 AG8010-0358 AG8010-0363 AG8010-0371 AG9301-0714 AGG4200-80119 AGG4513-60560 AGG4513-60561 AGG4513-80210 AGG4513-80211 AGG4513-80212 AGG4513-80218 AGG4513-80221 AGG4513-80222 AGG4513-80241 AGG4513-80242 AGG6500-80107 AGG6500-80114 AGG6500-80115 AG5182-9622 AG5182-9626 AG5182-9734 AG5188-6485 AG5190-3984 AG8010-0090 AG8010-0138 AG8010-0169 AG8010-0173 AG8010-0184 AG8010-0197 AG8010-0266 AG8010-0271 AG8010-0353 AG8010-0360 AG8010-0361 AG8010-0362 AGG4200-80118 AGG4513-80207 AGG4513-80219 AGG4513-80223 AGG4513-80224 AGG4513-80228 AGG4513-80230 AGG4513-80233 AGG6500-80111 AGG6500-80113
GC Autosampler Syringes, Agilent Technologies
Syringes Syringes for Chromatography
Agilent syringes are what you need for precise, effective, sample delivery from your GC autosampler.
  • Blue line syringes precisely match with your autosampler’s stroking mechanism for more accurate volume delivery
  • The smooth needle of the blue line syringes reduces septum coring and keeps your system working at its full potential
  • Blue line syringes are available in a choice of PTFE-tipped or metal fitted plunger to meet your application needs
  • Blue line syringes are delivered in easy-open, environmentally friendly packaging
  • Lot numbers are printed directly on the barrel of gold standard syringes, with a corresponding Certificate of Conformance ensuring certified performance to all specifications
  • The gold protective cap on the fused needle of the gold standard syringes prevents the glass syringe barrel from chipping as it is pressed against the inlet
  • Black ink and a gold illuminating backing strip enables effortless viewing of the volume scale of gold standard syringes and is easily discernible from imitators
  • Environmentally friendly packaging, with an improved design that reduces waste
  • Gold standard syringes are individually sealed, for contaminant-free use right out of the box
Agilent has what you need for precise, effective, sample delivery from your GC autosampler. Agilent offers two major lines of syringes for Agilent GC autosamplers: Gold standard and blue line syringes. Gold standard syringes are a versatile and economical choice suitable for most general applications, and are also available for Perkin Elmer, Shimadzu, Thermo, and Varian/Bruker GC systems. Blue line syringes are designed to support the higher productivity features of the Agilent 7693A automatic liquid sampler, while increasing plunger life and reducing costly downtime. Bulk purchasing options are available for both gold standard and blue line syringes.