Capillary Column Rinse Kit for GC, Agilent Technologies
Supplier: Avantor
AG430-3000KT 259.01 USD
AG430-3000
Capillary Column Rinse Kit for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent's capillary column rinse kit for GC systems makes solvent rinsing easy. Passing solvents through the column removes many contaminants that accumulate in it.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise.
This kit makes solvent rinsing easy! Solvent rinsing removes many contaminants that accumulate in columns. The contaminants are removed by passing solvents through the column. Only bonded and cross-linked phase columns should be rinsed, or permanent damage to the column will result.