DB-1 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
122-1012 122-1032 122-1033 122-1062 122-1063 123-1013 123-1015 123-1032 123-1033 123-1063 123-1064 125-1011 125-1032 125-1034 125-1035 128-1012 121-1022 121-1023 122-1013 122-106E 123-1012 123-1014 123-1032E 123-1033E 123-1034 123-1035 123-103B 123-1062 123-1065 125-1002E 125-1012 125-1015 125-101J 125-1035E 125-103J 125-1055 125-1065 126-1013 127-1022 G3903-61004 123-1011 123-1022 123-1026 123-1027 123-102F 123-1031 123-1032-INT 123-1033-INT 123-1034E 123-103E 123-1055 123-1056 123-105C 123-105F 123-1061 123-1063E 123-1065-INT 123-1065E 123-106B 123-106E 123-106G 124-1032 124-1034 125-1002 125-1005 125-100B 125-1011E 125-1014 125-1015E 125-1017 125-101K 125-1025 125-102J 125-1034E 125-1037 125-1039 125-103B 125-103K 125-103KE 125-1062 125-1064 125-106J 125-106JE 125-10B5 125-10H5 125-10HB 126-1012 127-100A 127-1012 127-1012E 127-1013 127-1013E 127-1023 127-1043 127-1046 127-1046E 128-1022 128-1052 12A-1015
AG122-1012EA 432 USD
AG122-1012 AG122-1032 AG122-1033 AG122-1062 AG122-1063 AG123-1013 AG123-1015 AG123-1032 AG123-1033 AG123-1063 AG123-1064 AG125-1011 AG125-1032 AG125-1034 AG125-1035 AG128-1012 AG121-1022 AG121-1023 AG122-1013 AG122-106E AG123-1012 AG123-1014 AG123-1032E AG123-1033E AG123-1034 AG123-1035 AG123-103B AG123-1062 AG123-1065 AG125-1002E AG125-1012 AG125-1015 AG125-101J AG125-1035E AG125-103J AG125-1055 AG125-1065 AG126-1013 AG127-1022 AGG3903-61004 AG123-1011 AG123-1022 AG123-1026 AG123-1027 AG123-102F AG123-1031 AG123-1032-INT AG123-1033-INT AG123-1034E AG123-103E AG123-1055 AG123-1056 AG123-105C AG123-105F AG123-1061 AG123-1063E AG123-1065-INT AG123-1065E AG123-106B AG123-106E AG123-106G AG124-1032 AG124-1034 AG125-1002 AG125-1005 AG125-100B AG125-1011E AG125-1014 AG125-1015E AG125-1017 AG125-101K AG125-1025 AG125-102J AG125-1034E AG125-1037 AG125-1039 AG125-103B AG125-103K AG125-103KE AG125-1062 AG125-1064 AG125-106J AG125-106JE AG125-10B5 AG125-10H5 AG125-10HB AG126-1012 AG127-100A AG127-1012 AG127-1012E AG127-1013 AG127-1013E AG127-1023 AG127-1043 AG127-1046 AG127-1046E AG128-1022 AG128-1052 AG12A-1015
DB-1 Columns, Agilent Technologies
Chromatography Columns
A reliable, crosslinked, nonpolar column with low bleed and a high temperature limit, delivering superb performance for general purpose applications.
  • 100% Dimethylpolysiloxane
  • Nonpolar and low bleed with a high temperature limit
  • Wide range of applications
  • Bonded, crosslinked, and solvent rinsible
  • Equivalent to USP Phase G2
  • Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1
  • Available with DuraGuard (GC column with a built-in guard column)
  • Wide range of column dimensions available
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-1 is nonpolar and low-bleed and operates under high temperature limits. It is an excellent choice for general purpose use in a wide variety of applications. Precision-engineered DB-1 is bonded, crosslinked and solvent rinsible, and is equivalent to USP phase G2.