DB-5 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
121-5022 122-5012 122-5032 122-5033 122-5062 122-5631 123-500ELTM 123-5032 123-5033 125-5032 127-5013 G3900-65006 121-5012 121-5012E 121-5042 122-5013 122-503E 122-5061 122-5063 122-5632 123-5012 123-5013 123-5032E 123-503B 123-503E 123-5056 125-5012 125-5035 125-503J 125-5065 127-501E 122-5002LTM 122-5011 122-501E 122-5022 122-5031 122-5032-INT 122-5032E 122-5033LTM 122-5052 122-5063E 123-5003LTM 123-500E 123-5011 123-5012LTM 123-5013E 123-5022 123-5026 123-502D 123-5031 123-5032-INT 123-5033E 123-5034 123-503ELTM 123-5053 123-5062 123-5063 124-5032 124-5037 125-5017 125-501J 125-501K 125-5025 125-5032E 125-5034 125-5035E 125-5037 125-503B 125-503D 125-503K 125-5062 125-5065E 125-50HB 127-5012 127-501N 127-5022 127-5023 128-5012 128-5022
AG121-5022EA 621 USD
AG121-5022 AG122-5012 AG122-5032 AG122-5033 AG122-5062 AG122-5631 AG123-500ELTM AG123-5032 AG123-5033 AG125-5032 AG127-5013 AGG3900-65006 AG121-5012 AG121-5012E AG121-5042 AG122-5013 AG122-503E AG122-5061 AG122-5063 AG122-5632 AG123-5012 AG123-5013 AG123-5032E AG123-503B AG123-503E AG123-5056 AG125-5012 AG125-5035 AG125-503J AG125-5065 AG127-501E AGG3903-63006 AG122-5002LTM AG122-5011 AG122-501E AG122-5022 AG122-5031 AG122-5032-INT AG122-5032E AG122-5033LTM AG122-5052 AG122-5063E AG123-5003LTM AG123-500E AG123-5011 AG123-5012LTM AG123-5013E AG123-5022 AG123-5026 AG123-502D AG123-5031 AG123-5032-INT AG123-5033E AG123-5034 AG123-503ELTM AG123-5053 AG123-5062 AG123-5063 AG124-5032 AG124-5037 AG125-5017 AG125-501J AG125-501K AG125-5025 AG125-5032E AG125-5034 AG125-5035E AG125-5037 AG125-503B AG125-503D AG125-503K AG125-5062 AG125-5065E AG125-50HB AG127-5012 AG127-501N AG127-5022 AG127-5023 AG128-5012 AG128-5022
DB-5 Columns, Agilent Technologies
Chromatography Columns
High-quality, nonpolar, general-purpose columns with low bleed and a high temperature limit.
  • (5%-Phenyl)-methylpolysiloxane
  • Nonpolar, low bleed and with a high temperature limit
  • Bonded, crosslinked, and solvent rinsible
  • Wide range of column dimensions available
  • Excellent general purpose column with a wide range of applications
  • Equivalent to USP Phase G27
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-5 is nonpolar and low bleed and is available in a range of column dimensions. DB-5 is also bonded, crosslinked, and solvent-rinsible, and has a high temperature limit. It is equivalent to USP phase G27. The DB-5 column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency.