CrossLab Rotating Cylinder for Dissolution, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
12-1370
AG12-1370EA 624 USD
AG12-1370
CrossLab Rotating Cylinder for Dissolution, Agilent Technologies
Dissolution Apparatus
Uses a stainless steel cylinder and includes two sizes for smaller and larger patches.
  • No need to match an upper and lower assembly
The rotating cylinder for USP Apparatus 6 assembly uses a stainless steel cylinder and includes four slots at the top for improved media circulation within the dissolution vessel. Each rotating cylinder includes two sizes, one for smaller patches, the other for larger ones.